Chemical Identifiers
CAS62625-29-0
IUPAC Namesodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzen-1-olate
Molecular FormulaC21H17NaO5S
InChI KeyFMKJVYROEYMYFV-UHFFFAOYNA-M
SMILES[Na+].CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C(C)=C1
View more