Penicillin-Streptomycin (10,000 U/mL)
Save up to 29% on your order of cell culture products. View promotion details >
Try new Pen-Strep antibiotic with GlutaMAX supplement, a stable alternative to L-glutamine that enhances cell health. Learn more >
Penicillin-Streptomycin (10,000 U/mL)
Penicillin-Streptomycin (10,000 U/mL)
Penicillin-Streptomycin (10,000 U/mL)
Gibco™

Penicillin-Streptomycin (10,000 U/mL)

This solution contains 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin. The antibiotics penicillin and streptomycin are used toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
15140148
also known as 15140-148
20 mL
15140122
also known as 15140-122
100 mL
15140163
also known as 15140-163
20 x 100 mL
Catalog number 15140148
also known as 15140-148
Price (USD)
17.47
Each
Add to cart
Quantity:
20 mL
Price (USD)
17.47
Each
Add to cart
This solution contains 10,000 units/mL of penicillin and 10,000 μg/mL of streptomycin. The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Penicillin was originally purified from the fungi Penicillium and acts by interfering directly with the turnover of the bacterial cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to the 30S subunit of the bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.

We offer a wide range of antibiotics and antimycotics in both powder and liquid formats. Learn more about the following types of products:

Cell Culture Antibiotics
Selection Antibiotics (including recommended working concentrations)

Learn more about the use of antibiotics and antimycotics in cell culture and review guidelines for decontaminating cultures.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration100 X
For Use With (Application)Prevention of Cell Culture Contamination
Quantity20 mL
Shelf Life12 Months
Shipping ConditionDry Ice
FormLiquid
Product TypeAntibiotic
SterilitySterile-filtered
Sterilization MethodSterile-filtered
Unit SizeEach
Contents & Storage
Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

My Penicillin-Streptomycin solution is not colorless. Is this normal?

Yes, this is normal and will not affect the potency or application of the product. This solution is typically colorless. However, it can have a pink to yellow color tint. The coloring is a carry-over from the manufacturing process of Streptomycin - the genus that Steptomycin is isolated from (Actinomycetes Streptomyces griseus) is responsible for a wide variety of pigments.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How can I decontaminate my cultures?

When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.

1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.

The following is a suggested procedure for determining toxicity levels and decontaminating cultures:

1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What antibiotics do you offer to help control or eliminate cell culture contamination?

Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Share catalog number, name or link