Thermo Scientific Chemicals

Methyl Red, sodium salt, indicator, pure

Catalog number: 151415000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl Red, sodium salt, indicator, pure

Catalog number: 151415000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151415000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
845-10-3
IUPAC Name
sodium 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular Formula
C15H14N3NaO2
InChI Key
GNTPCYMJCJNRQB-UHFFFAOYSA-M
SMILES
[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
Appearance (Color)
Orange to red to brown
Infrared spectrum
Conforms
Visual transition interval
From pH 4.2 (pink) to pH 6.2 (yellow)
Appearance (Form)
Powder
Solubility
(0.1% in water) clear to faint turbid solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text