Thermo Scientific Chemicals

Thymolphthalein, Indicator, Pure

Catalog number: 151460100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Thymolphthalein, Indicator, Pure

Catalog number: 151460100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151460100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
125-20-2
IUPAC Name
3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C28H30O4
InChI Key
LDKDGDIWEUUXSH-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
Appearance (Color)
White
Infrared spectrum
Conforms
Loss on drying
=<1 %
Appearance (Form)
Crystalline powder
Sulfated ash
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text