Thermo Scientific Chemicals

5-(4-Dimethylaminobenzylidene)rhodanine, 99%

Catalog number: 151471000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

5-(4-Dimethylaminobenzylidene)rhodanine, 99%

Catalog number: 151471000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151471000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
536-17-4
IUPAC Name
(5Z)-5-{[4-(dimethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Formula
C12H12N2OS2
InChI Key
JJRVRELEASDUMY-YFHOEESVSA-N
SMILES
CN(C)C1=CC=C(C=C2/SC(=S)NC2=O)C=C1
Appearance (Form)
Powder
Solubility
(0.03 % in acetone) Clear
Titration with TBAH
98.5 to 101.5 %
Infrared spectrum
Conforms
Suitability
(for Ag determination) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text