Isatin, 98%, Thermo Scientific Chemicals
Isatin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isatin, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151495000
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
Add to cart
Isatin, 98%, Thermo Scientific Chemicals
Catalog number151495000
Price (USD)/ Each
147.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
174.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H5NO2
InChI KeyJXDYKVIHCLTXOP-UHFFFAOYSA-N
SMILESO=C1NC2=CC=CC=C2C1=O
Molecular Weight (g/mol)147.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point199°C to 204°C
Water=<1.0 % (K.F.)
Infrared spectrumConforms
Appearance (Color)Orange to red
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only