Thermo Scientific Chemicals

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog number: 151531000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog number: 151531000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151531000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
131-91-9
IUPAC Name
1-nitrosonaphthalen-2-ol
Molecular Formula
C10H7NO2
InChI Key
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC=CC2=C1N=O
Water
=<0.5 %
Appearance (Color)
Brown
Infrared spectrum
Conforms
Titration with NaOH
97.5 to 102.5 % (On dry substance)
Melting point
104°C to 108°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text