Chemical Identifiers
CAS23647-14-5
IUPAC Nametrisodium (3E)-5-hydroxy-4-oxo-3-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC16H9N2Na3O11S3
InChI KeyLSAWBVPBZOEGOT-ZOVWSQMSSA-K
SMILES[Na+].[Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)S([O-])(=O)=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
View more