Thermo Scientific Chemicals

2-(4-Sulfophenylazo)1,8-dihydroxy-3,6-naphthalenedisulfonic acid, trisodium salt, in

Catalog number: 151571000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(4-Sulfophenylazo)1,8-dihydroxy-3,6-naphthalenedisulfonic acid, trisodium salt, in

Catalog number: 151571000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151571000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23647-14-5
IUPAC Name
trisodium (3E)-5-hydroxy-4-oxo-3-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Molecular Formula
C16H9N2Na3O11S3
InChI Key
LSAWBVPBZOEGOT-ZOVWSQMSSA-K
SMILES
[Na+].[Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)S([O-])(=O)=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Appearance (Color)
Dark red-brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
UV spectrum
Complies

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text