Thermo Scientific Chemicals

Tetrabromophenolphthalein Ethyl Ester, Potassium Salt, Indicator Grade, Thermo Scientific Chemicals

Catalog number: 151580010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrabromophenolphthalein Ethyl Ester, Potassium Salt, Indicator Grade, Thermo Scientific Chemicals

Catalog number: 151580010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151580010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62637-91-6
IUPAC Name
potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
Molecular Formula
C22H13Br4KO4
InChI Key
WCIQBKUTYDIBJC-UHFFFAOYSA-M
SMILES
[K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
Specific extinction E (1%/1cm)
>=660 (at lambda max, in ethanol)
Appearance (Color)
Dark green to dark blue to black
Infrared spectrum
Conforms
Sulfated ash
10 % to 15 %
UV Lambda max
600 to 608 nm (in ethanol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text