Myricetin, 95%, Thermo Scientific Chemicals
Myricetin, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Myricetin, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 mg
Packaging:
Glass bottle
Catalog number 151590250
Price (USD)/ Each
-
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
Molecular FormulaC15H10O8
InChI KeyIKMDFBPHZNJCSN-UHFFFAOYSA-N
SMILESOC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1
Molecular Weight (g/mol)318.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown-yellow to green-brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only