Thermo Scientific Chemicals

4,4'-Difluorobenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: 151790250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4,4'-Difluorobenzophenone, 99%, Thermo Scientific Chemicals

Catalog number: 151790250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151790250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
345-92-6
IUPAC Name
bis(4-fluorophenyl)methanone
Molecular Formula
C13H8F2O
InChI Key
LSQARZALBDFYQZ-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
Appearance (Form)
Crystalline powder
Melting point
106°C to 109°C
Loss on drying
=<0.25 %
Appearance (Color)
White to light yellow
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text