Thermo Scientific Chemicals

L(-)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals

Catalog number: 151830250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

L(-)-alpha-Methylbenzylamine, 99+%, (99% ee), Thermo Scientific Chemicals

Catalog number: 151830250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151830250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2627-86-3
IUPAC Name
(1S)-1-phenylethan-1-amine
Molecular Formula
C8H11N
InChI Key
RQEUFEKYXDPUSK-ZETCQYMHSA-N
SMILES
C[C@H](N)C1=CC=CC=C1
Specific optical rotation
-39° to -41° (20°C, 589 nm) (neat)
Enantiomeric excess
>=98.5 % (GC)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text