Hexanal, 96%, stabilized, Thermo Scientific Chemicals
Hexanal, 96%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexanal, 96%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 151850010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS66-25-1
IUPAC Namehexanal
Molecular FormulaC6H12O
InChI KeyJARKCYVAAOWBJS-UHFFFAOYSA-N
SMILESCCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Acid value=<10 mg KOH/g
Density(20°C) 0.810 to 0.820 g/ml
GC>=95 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only