Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Binding Property | Standard Capacity, Affinity |
Description | Clear, 96-well |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein L |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 8-well Strip |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Detection Plate, Immunoassay, ELISA |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein A |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 96-well |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein A |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Black, 96-well |
Detection Method | Fluorescence |
Product Line | Pierce |
Color | Black |
Product Type | Detection Plate, Immunoassay, ELISA |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein A |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | White, 96-well |
Detection Method | Chemiluminescence |
Product Line | Pierce |
Color | White |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein A |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 8-well Strip |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein G |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | Each |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 96-well |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Detection Plate, Immunoassay, ELISA |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein G |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | Each |
Binding Property | Standard Capacity, Affinity |
Description | Black, 96-well |
Detection Method | Fluorescence |
Product Line | Pierce |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein G |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | Each |
Binding Property | Standard Capacity, Affinity |
Description | White, 96-well |
Detection Method | Chemiluminescence |
Product Line | Pierce |
Color | White |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein G |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | Each |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 8-well Strip |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Detection Plate, Immunoassay, ELISA |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Protein A/G |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Catalog Number | Specifications | Unit Size | Surface Treatment | Description | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
15190 | 5 plates | Protein L | Clear, 96-well | 288.65 Online price 300.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15132 | 5 plates | Protein A | Clear, 8-well Strip | 268.65 Online price 279.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15130 | 5 plates | Protein A | Clear, 96-well | 236.65 Online price 246.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15155 | 5 plates | Protein A | Black, 96-well | 268.65 Online price 279.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15154 | 5 plates | Protein A | White, 96-well | 268.65 Online price 279.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15133 | Each | Protein G | Clear, 8-well Strip | 291.65 Online price 303.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15131 | Each | Protein G | Clear, 96-well | 268.65 Online price 279.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15157 | Each | Protein G | Black, 96-well | 291.65 Online price 303.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15156 | Each | Protein G | White, 96-well | 291.65 Online price 303.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15138 | 5 plates | Protein A/G | Clear, 8-well Strip | 294.65 Online price 306.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific Pierce Protein Coated Plates provide an alternative to direct, passive adsorption methods for immobilizing antibodies for ELISA and other plate-based assay techniques.
Highlights
• Retain antibody activity, which can be lost when antibodies are immobilized by passive adsorption
• Orient antibodies for maximum antigen-binding capacity
• Immobilize antibodies for plate assays without prior purification
• Ensure minimal variation (<5% well-to-well) from consistent coating
• Reduce nonspecific binding because plates are pre-blocked with SuperBlock Blocking Buffer
• Bind 2 to 5 pmol of antibody per well (based on tests with rabbit IgG)
Protein A/G Coated Plates
• Fusion protein containing four antibody-binding sites from Protein A and two from Protein G
• Binds to most IgG species because it combines the specificities of Protein A and Protein G
• Binds to Fc region of antibodies, ensuring optimal orientation
• Binds well using a wide range of pH
Protein A Coated Plates
• Binds strongly to IgG from human, rabbit, guinea pig, pig, dog, and rhesus monkey
• Binds strongly to mouse IgG2a, IgG2b and IgG3
• Binds to Fc region of antibodies for optimal orientation
Protein G Coated Plates
• Binds strongly to IgG from many species including human, mouse, rabbit, sheep and goat
• Binds only to IgG—no cross-reactivity with other antibody classes
• Binds to Fc region of antibodies for optimal orientation
Protein L Coated Plates
• Binds to all classes of immunoglobulins (IgG, IgM, IgA, IgE and IgD)
• Binds to the VL region of kappa light chains (human I, III, IV and Mouse I) without interfering with antigen-binding sites
• Binds to Fab fragments and ScFv
• Does not bind bovine, goat or sheep immunoglobulin
• Binds weakly to rabbit immunoglobulin
• Pre-blocked to reduce nonspecific binding