Thermo Scientific Chemicals

2,2,6,6-Tetramethyl-4-piperidinol, 99%, Thermo Scientific Chemicals

Catalog number: 151942500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2,6,6-Tetramethyl-4-piperidinol, 99%, Thermo Scientific Chemicals

Catalog number: 151942500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 151942500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2403-88-5
IUPAC Name
2,2,6,6-tetramethylpiperidin-4-ol
Molecular Formula
C9H19NO
InChI Key
VDVUCLWJZJHFAV-UHFFFAOYSA-N
SMILES
CC1(C)CC(O)CC(C)(C)N1
Infrared spectrum
Conforms
Appearance (Color)
White to off-white to light beige
Titration with HClO4
>=98.5 %
Appearance (Form)
Crystalline powder or crystals and/or chunks
Melting point
129°C to 133°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text