96 Well Black/Clear Bottom Plate, Collagen I Coated Surface, Pack of 5
Tired of paying too much for ECM plates? Try these ECM plates that are easy on your budget »
96 Well Black/Clear Bottom Plate, Collagen I Coated Surface, Pack of 5
Thermo Scientific™
96 Well Black/Clear Bottom Plate, Collagen I Coated Surface, Pack of 5
Color:Black, Clear Bottom; Lid:With Lid; Type: Optical Polymer Base
Have Questions?
Catalog number 152036
Price (USD)/ Case of 20
626.00
-
Add to cart
Price (USD)/ Case of 20
626.00
Add to cart
96 Well Black/Clear Bottom Plate, Collagen I Coated Surface, Pack of 5
Catalog number152036
Price (USD)/ Case of 20
626.00
-
Add to cart
Thermo Scientific™ Nunc MicroWell 96-Well Optical-Bottom Plates with Polymer Base have a solid polystyrene black or white upper structures with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.
White or black polystyrene upper structure with polymer or coverglass bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.
  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Available in black for fluorescence assays or white for luminescence and fluorescence assays; both with clear bottoms
  • Polymer base plate
  • Working range, 50-200μL/well
  • Customized bar coding is available
  • Aseptically manufactured
Ordering Information:
Part No. 152040, 152036, 152028 and 152037 is produced in a clean environment.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
MaterialPolystyrene
Volume (Metric) Well400 μL
Volume (Metric) Working50 to 200 μL
TypeOptical Polymer Base
QuantityCase of 20
No. per Pack5
No. per Case20
ColorWhite
Surface TreatmentCollagen Type I
SterilityAseptically Manufactured
No. of Wells96
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Array8 x 12
Product LineNunc™
Packaging5/Pack, 20/Case
BarcodeCustom Barcoding Available
Well ShapeFlat
Unit SizeCase of 20