Nunc™ MicroWell™ 96-Well, Collagen Type I-Treated, Flat-Bottom, Optical Polymer Base Microplate
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ MicroWell™ 96-Well, Collagen Type I-Treated, Flat-Bottom, Optical Polymer Base Microplate
Thermo Scientific™
Nunc™ MicroWell™ 96-Well, Collagen Type I-Treated, Flat-Bottom, Optical Polymer Base Microplate
Color:White, Clear Bottom; Lid:With Lid; Type: Optical Polymer Base
Have Questions?
Catalog NumberQuantity
152040Caseof 20
Catalog number 152040
Price (USD)
738.00
Case of 20
Add to cart
Quantity:
Caseof 20
Price (USD)
738.00
Case of 20
Add to cart
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Polymer Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Customized bar coding is available
  • Aseptically manufactured
Ordering Information:
Part No. 152040, 152036, 152028 and 152037 is produced in a clean environment.
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Array8 x 12
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
ColorClear Bottom
No. per Case20
No. per Pack5
Packaging5 Pk., 20 Cases
Product LineNunc™
QuantityCaseof 20
Surface TreatmentCollagen Type I
Well ShapeFlat
MaterialPolystyrene
No. of Wells96
SterilityAseptically Manufactured
TypeMicroplate
Volume (Metric) Well400 μL
Unit SizeCase of 20
Frequently asked questions (FAQs)
What does it mean for a plate to be "Aseptically Manufactured"?
Prior to treatment with an extra cellular matrix (ECM), the plates are sterilized. The ECM treatment is a process that utilizes sterile technique, but we cannot claim the product is "sterile" because it has been opened. If the product were to be re-sterilized after the treatment, the ECM coating would be ruined. For the purpose of using this plate in cell-culture applications, it is functionally sterile.
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link