Thermo Scientific Chemicals

N-Piperidineethanol, 99%, Thermo Scientific Chemicals

Catalog number: 152115000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Piperidineethanol, 99%, Thermo Scientific Chemicals

Catalog number: 152115000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 152115000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3040-44-6
IUPAC Name
2-(piperidin-1-yl)ethan-1-ol
Molecular Formula
C7H15NO
InChI Key
KZTWONRVIPPDKH-UHFFFAOYSA-N
SMILES
OCCN1CCCCC1
Appearance (Color)
Clear light brown
GC
>=98.5 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4794 to 1.4814 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text