Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
1 kg
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 152130100
Price (USD)/ Each
575.65
Online exclusive
639.00 
Save 63.35 (10%)
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
575.65
Online exclusive
639.00 
Save 63.35 (10%)
Add to cart
Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
Catalog number152130100
Price (USD)/ Each
575.65
Online exclusive
639.00 
Save 63.35 (10%)
-
Add to cart
Chemical Identifiers
CAS76-03-9
IUPAC Nametrichloroacetic acid
Molecular FormulaC2HCl3O2
InChI KeyYNJBWRMUSHSURL-UHFFFAOYSA-N
SMILESOC(=O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Iron (Fe)=<10 ppm
Appearance (Form)Crystalline powder or crystals or flakes
Sulfuric acid=<0.3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only