Diethyl vinylphosphonate, 97%, Thermo Scientific Chemicals
Diethyl vinylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl vinylphosphonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
Packaging:
Glass bottle
Catalog number 152150250
Price (USD)/ Each
255.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
301.00 
Save 45.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
255.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
301.00 
Save 45.35 (15%)
Add to cart
Diethyl vinylphosphonate, 97%, Thermo Scientific Chemicals
Catalog number152150250
Price (USD)/ Each
255.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
301.00 
Save 45.35 (15%)
-
Add to cart
Chemical Identifiers
CAS682-30-4
IUPAC Namediethyl ethenylphosphonate
Molecular FormulaC6H13O3P
InChI KeyDREPONDJUKIQLX-UHFFFAOYSA-N
SMILESCCOP(=O)(OCC)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4280 to 1.4300 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only