Thermo Scientific Chemicals

1-Benzylimidazole, 99%, Thermo Scientific Chemicals

Catalog number: 152250250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Benzylimidazole, 99%, Thermo Scientific Chemicals

Catalog number: 152250250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 152250250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4238-71-5
IUPAC Name
1-benzyl-1H-imidazole
Molecular Formula
C10H10N2
InChI Key
KKKDZZRICRFGSD-UHFFFAOYSA-N
SMILES
C(N1C=CN=C1)C1=CC=CC=C1
Appearance (Color)
White to brown
Appearance (Form)
Crystalline powder or crystals
Melting point
68°C to 72°C
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text