Gibco™

Distilled Water Bag(s)

Catalog number: 15230253
Have Questions?
Gibco™

Distilled Water Bag(s)

Catalog number: 15230253
Have Questions?
Catalog Number
15230253
Unit Size
5 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
15230279
also known as 15230-279
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
152302535 L
-
1523026110 L
-
15230279
also known as 15230-279
20 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, this distilled water features:

• Cell culture quality
• Proven performance
• Suitable for use in cell culture and molecular biology applications

Cell culture quality
This water is prepared by distillation and filtration (0.1 μm) with no added substances. Multiple aspects of water quality are monitored on a routine basis: conductivity, total organic carbon (TOC), bioburden, endotoxin, and metals.

Proven performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.

Use in cell culture and molecular biology applications
This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

pH
6 to 8
Recommended Storage
Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Grade
Cell Culture
Packaging
Universal Bag
Sterility
Sterile-filtered
Treatment(s)
Not DEPC-Treated
Quantity
5 L
Purification Method
Membrane-Filtered
Shipping Condition
Room Temperature
Product Line
Gibco™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text