Thermo Scientific Chemicals

(2-Bromoethyl)trimethylammonium Bromide 98%, Thermo Scientific Chemicals

Catalog number: 152350250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

(2-Bromoethyl)trimethylammonium Bromide 98%, Thermo Scientific Chemicals

Catalog number: 152350250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 152350250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2758-06-7
IUPAC Name
(2-bromoethyl)trimethylazanium bromide
Molecular Formula
C5H13Br2N
InChI Key
OINMNSFDYTYXEQ-UHFFFAOYSA-M
SMILES
[Br-].C[N+](C)(C)CCBr
Appearance (Color)
Light beige to light brown
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 %
Melting point
240°C to 244°C
Appearance (Form)
Powder and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text