4'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals
4'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 152360010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS100-06-1
IUPAC Name1-(4-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O2
InChI KeyNTPLXRHDUXRPNE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline mass
Melting point36°C to 40°C
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks or
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only