1x1 image pixel for data collection
p-Anisic acid, 98%
p-Anisic acid, 98%
p-Anisic acid, 98%
Thermo Scientific Chemicals

p-Anisic acid, 98%

CAS: 100-09-4 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 kg
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 152371000
Price (USD)
44.65
Online Exclusive
49.00
Save 4.35 (9%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.00
Save 4.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS100-09-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point181°C to 186°C
Infrared spectrumConforms
Appearance (Color)White to beige to light grey
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link