Thermo Scientific Chemicals

3-Aminocrotononitrile, 96%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 152451000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3-Aminocrotononitrile, 96%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 152451000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 152451000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1118-61-2
IUPAC Name
(2E)-3-aminobut-2-enenitrile
Molecular Formula
C4H6N2
InChI Key
DELJOESCKJGFML-DUXPYHPUSA-N
SMILES
C\C(N)=C/C#N
Appearance (Color)
Yellow
Infrared spectrum
Conforms
Appearance (Form)
Flakes
GC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text