Phenylacetylene, 98%, pure
Phenylacetylene, 98%, pure
Phenylacetylene, 98%, pure
Phenylacetylene, 98%, pure
Thermo Scientific Chemicals

Phenylacetylene, 98%, pure

CAS: 536-74-3 | C8H6 | 102.14 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 152460250
Price (JPY)
-
Request A Quote
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS536-74-3
IUPAC Nameethynylbenzene
Molecular FormulaC8H6
InChI KeyUEXCJVNBTNXOEH-UHFFFAOYSA-N
SMILESC#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5470 to 1.5500 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link