Thermo Scientific Chemicals

m-Anisic acid, 98%, Thermo Scientific Chemicals

Catalog number: 152471000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

m-Anisic acid, 98%, Thermo Scientific Chemicals

Catalog number: 152471000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 152471000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
586-38-9
IUPAC Name
3-methoxybenzoic acid
Molecular Formula
C8H8O3
InChI Key
XHQZJYCNDZAGLW-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)C(O)=O
Appearance (Form)
Powder
GC
>=97.5 %
Appearance (Color)
Off-white to yellow to light beige
Infrared spectrum
Conforms
Melting point
103°C to 108°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text