1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals
1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Adamantanecarboxylic acid chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 152480250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameadamantane-1-carbonyl chloride
Molecular FormulaC11H15ClO
InChI KeyMIBQYWIOHFTKHD-UHFFFAOYSA-N
SMILESClC(=O)C12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol)198.69
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point48°C to 52°C
Appearance (Form)Crystalline solid
Titration RCOCl>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only