Isopropenyl acetate, 99%, Thermo Scientific Chemicals
Isopropenyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropenyl acetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 152502500
Price (USD)/ Each
29.00
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
29.00
Add to cart
Isopropenyl acetate, 99%, Thermo Scientific Chemicals
Catalog number152502500
Price (USD)/ Each
29.00
-
Add to cart
Chemical Identifiers
CAS108-22-5
IUPAC Nameprop-1-en-2-yl acetate
Molecular FormulaC5H8O2
InChI KeyHETCEOQFVDFGSY-UHFFFAOYSA-N
SMILESCC(=C)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.3990 to 1.4030 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only