Thermo Scientific Chemicals

trans-Anethole, 99%, Thermo Scientific Chemicals

Catalog number: 152535000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

trans-Anethole, 99%, Thermo Scientific Chemicals

Catalog number: 152535000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 152535000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4180-23-8
IUPAC Name
1-methoxy-4-[(1E)-prop-1-en-1-yl]benzene
Molecular Formula
C10H12O
InChI Key
RUVINXPYWBROJD-ONEGZZNKSA-N
SMILES
COC1=CC=C(\C=C\C)C=C1
Density
(25/4°C) 0.980 to 0.985
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.5580 to 1.5650 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text