Thermo Scientific Chemicals

Rhodanine 99%, Thermo Scientific Chemicals

Catalog number: 152601000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Rhodanine 99%, Thermo Scientific Chemicals

Catalog number: 152601000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 152601000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-84-4
IUPAC Name
2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Formula
C3H3NOS2
InChI Key
KIWUVOGUEXMXSV-UHFFFAOYSA-N
SMILES
O=C1CSC(=S)N1
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
167°C to 170°C
Titration with NaOH
>=98.5 %
Appearance (Color)
Light yellow to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text