9-Decen-1-ol, 99%, Thermo Scientific Chemicals
9-Decen-1-ol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9-Decen-1-ol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 152640050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13019-22-2
IUPAC Namedec-9-en-1-ol
Molecular FormulaC10H20O
InChI KeyQGFSQVPRCWJZQK-UHFFFAOYSA-N
SMILESOCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4460 to 1.4500 (20°C, 589 nm)
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only