3,3-Dimethylpentane, 99%, Thermo Scientific Chemicals
3,3-Dimethylpentane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethylpentane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 152660050
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
Add to cart
3,3-Dimethylpentane, 99%, Thermo Scientific Chemicals
Catalog number152660050
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS562-49-2
IUPAC Name3,3-dimethylpentane
Molecular FormulaC7H16
InChI KeyAEXMKKGTQYQZCS-UHFFFAOYSA-N
SMILESCCC(C)(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.3900 to 1.3920 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only