3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
Thermo Scientific Chemicals

3,3-Dimethylpentane, 99%

CAS: 562-49-2 | C7H16 | 100.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 152660250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS562-49-2
IUPAC Name3,3-dimethylpentane
Molecular FormulaC7H16
InChI KeyAEXMKKGTQYQZCS-UHFFFAOYSA-N
SMILESCCC(C)(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
Refractive index1.3900 to 1.3920 (20°C, 589 nm)
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471235Certificate of AnalysisMay 17, 2025152660010, 152660050, 152660250
A0472215Certificate of AnalysisMay 17, 2025152660010, 152660050, 152660250
A0429966Certificate of AnalysisNov 13, 2023152660010, 152660050, 152660250
A0450517Certificate of AnalysisMay 02, 2023152660010, 152660050, 152660250
A0392457Certificate of AnalysisDec 15, 2022152660010, 152660050, 152660250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection