2,4,6-Triisopropylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
2,4,6-Triisopropylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Triisopropylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 152790250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2,4,6-tris(propan-2-yl)benzene-1-sulfonyl chloride
Molecular FormulaC15H23ClO2S
InChI KeyJAPYIBBSTJFDAK-UHFFFAOYSA-N
SMILESCC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(Cl)(=O)=O
Molecular Weight (g/mol)302.86
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline solid
Melting point93°C to 97°C
Titration Argentometric>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only