Thermo Scientific Chemicals

2,4,6-Triisopropylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 152791000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4,6-Triisopropylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 152791000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 152791000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6553-96-4
IUPAC Name
2,4,6-tris(propan-2-yl)benzene-1-sulfonyl chloride
Molecular Formula
C15H23ClO2S
InChI Key
JAPYIBBSTJFDAK-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(Cl)(=O)=O
Titration Argentometric
>=96.0 %
Appearance (Form)
Crystalline solid
Melting point
93°C to 97°C
Appearance (Color)
White to light beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text