2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals
2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 152870100
Price (USD)/ Each
102.65
Online exclusive
121.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
121.00 
Save 18.35 (15%)
Add to cart
2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals
Catalog number152870100
Price (USD)/ Each
102.65
Online exclusive
121.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Molecular FormulaC12H6Cl2NNaO2
InChI KeyCVSUAFOWIXUYQA-UHFFFAOYSA-M
SMILES[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
Molecular Weight (g/mol)290.07
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark blue-purple
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO494 to 106 % (On dry substance)
Loss on drying=<14 % (1 g, 120°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only