Chemical Identifiers
CAS17372-87-1
IUPAC Namedisodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6Br4Na2O5
InChI KeySEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
View more