Thermo Scientific Chemicals

4-Morpholinoacetophenone, 99%, Thermo Scientific Chemicals

Catalog number: 152900500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Morpholinoacetophenone, 99%, Thermo Scientific Chemicals

Catalog number: 152900500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 152900500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39910-98-0
IUPAC Name
1-[4-(morpholin-4-yl)phenyl]ethan-1-one
Molecular Formula
C12H15NO2
InChI Key
AKQWEDMTPCAESO-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(C=C1)N1CCOCC1
Appearance (Form)
Crystalline powder
GC
>=98.5 %
Appearance (Color)
Yellow to light brown
Infrared spectrum
Conforms
Melting point
94°C to 98°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text