Thermo Scientific Chemicals

Amaranth, 85%

Catalog number: 153031000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Amaranth, 85%

Catalog number: 153031000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153031000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
915-67-3
IUPAC Name
trisodium (4E)-3-oxo-4-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Molecular Formula
C20H11N2Na3O10S3
InChI Key
VOBHRQFELWTZFS-DXVNCNPQSA-K
SMILES
[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N\NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O
Appearance (Form)
Crystalline powder
UV spectrophotometry
>=80 %
Appearance (Color)
Dark red-brown to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text