Chemical Identifiers
CAS915-67-3
IUPAC Nametrisodium (4E)-3-oxo-4-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC20H11N2Na3O10S3
InChI KeyVOBHRQFELWTZFS-DXVNCNPQSA-K
SMILES[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N\NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O
View more