Thermo Scientific Chemicals

2-Fluorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 153091000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Fluorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 153091000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 153091000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
393-52-2
IUPAC Name
2-fluorobenzoyl chloride
Molecular Formula
C7H4ClFO
InChI Key
RAAGZOYMEQDCTD-UHFFFAOYSA-N
SMILES
FC1=CC=CC=C1C(Cl)=O
Appearance (Color)
Clear colorless to slightly colored
GC
>=96.0 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5355 to 1.5375 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text