Thermo Scientific Chemicals

3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 153191000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 153191000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 153191000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
99-34-3
IUPAC Name
3,5-dinitrobenzoic acid
Molecular Formula
C7H4N2O6
InChI Key
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Titration with NaOH
98.0 to 102.0 %
HPLC
>=98.0 %
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text