3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 153195000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3,5-dinitrobenzoic acid
Molecular FormulaC7H4N2O6
InChI KeyVYWYYJYRVSBHJQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Molecular Weight (g/mol)212.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point204°C to 209°C
Titration with NaOH98.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only