Thermo Scientific Chemicals

Fast Garnet GBC Base 97%, Thermo Scientific Chemicals

Catalog number: 153260250
25 g, Each
Thermo Scientific Chemicals

Fast Garnet GBC Base 97%, Thermo Scientific Chemicals

Catalog number: 153260250
25 g, Each
Quantity
Catalog number: 153260250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-56-3
IUPAC Name
2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline
Molecular Formula
C14H15N3
InChI Key
PFRYFZZSECNQOL-UHFFFAOYSA-N
SMILES
CC1=CC(=CC=C1N)N=NC1=CC=CC=C1C
Appearance (Form)
Powder
Appearance (Color)
Orange to red-brown to dark brown
Infrared spectrum
Conforms
Melting point
100°C to 104°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text