2-Deoxy-D-ribose, 99%
2-Deoxy-D-ribose, 99%
Thermo Scientific Chemicals

2-Deoxy-D-ribose, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 153280050
Price (USD)/ Each
72.70
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
72.70
Add to cart
2-Deoxy-D-ribose, 99%
Catalog number153280050
Price (USD)/ Each
72.70
-
Add to cart
Chemical Identifiers
IUPAC Name(4S,5R)-5-(hydroxymethyl)oxolane-2,4-diol
Molecular FormulaC5H10O4
InChI KeyPDWIQYODPROSQH-GLULIZFNNA-N
SMILESOC[C@H]1OC(O)C[C@@H]1O
Molecular Weight (g/mol)134.13
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Infrared spectrumConforms
Assay>=98.5 %
Loss on drying=<1.0 % (40°C, 2 hrs) (vacuum)
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only