Thermo Scientific Chemicals

2-Deoxy-D-ribose, 99%

Catalog number: 153280250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Deoxy-D-ribose, 99%

Catalog number: 153280250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 153280250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
533-67-5
IUPAC Name
(4S,5R)-5-(hydroxymethyl)oxolane-2,4-diol
Molecular Formula
C5H10O4
InChI Key
PDWIQYODPROSQH-GLULIZFNNA-N
SMILES
OC[C@H]1OC(O)C[C@@H]1O
Infrared spectrum
Conforms
Assay
>=98.5 %
Loss on drying
=<1.0 % (40°C, 2 hrs) (vacuum)
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text