Thermo Scientific Chemicals

Neocuproine Hydrochloride Monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 153310250
25 g, Each
Thermo Scientific Chemicals

Neocuproine Hydrochloride Monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 153310250
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 153310250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
303136-82-5
IUPAC Name
2,9-dimethyl-1,10-phenanthroline
Molecular Formula
C14H12N2
InChI Key
IYRGXJIJGHOCFS-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
Appearance (Color)
White to light yellow to beige
Appearance (Form)
Powder or crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 %
Water
5.0 to 10.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text